Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145099.2 | NP_659567.2 | 7 - 8 | 854 | 82 | |
GenBank mRNA | AB083654.1 | - | 7 - 8 | 830 | 82 | |
AF510316.1 | - | 7 - 8 | 854 | 82 | ||
BC108984.1 | - | 7 - 8 | 840 | 40047664 | 82 | |
BC108985.1 | - | 7 - 8 | 840 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |