Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145209.3 | NP_660210.1 | 2 - 3 | 293 | 53 | |
GenBank mRNA | AB067533.1 | - | 1 - 2 | 218 | 53 | |
AF426289.1 | - | 1 - 2 | 224 | 53 | ||
AK078690.1 | - | 2 - 3 | 294 | 53 | ||
AK089111.1 | - | 1 - 2 | 263 | 53 | ||
AK149824.1 | - | 1 - 2 | 217 | 53 | ||
AK150863.1 | - | 1 - 2 | 347 | 53 | ||
AK165578.1 | - | 1 - 2 | 330 | 53 | ||
AY057107.1 | - | 1 - 2 | 206 | 53 | ||
AY089728.1 | - | 1 - 2 | 218 | 53 | ||
BC032152.1 | - | 1 - 2 | 230 | 5068699 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |