Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282071.2 | NP_001269000.1 | 1 - 2 | 79 | 74 | |
NM_147779.2 | NP_680088.1 | 1 - 2 | 79 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |