Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148930.3 | NP_683732.1 | 21 - 22 | 2185 | 96 | |
GenBank mRNA | AJ309168.1 | - | 20 - 21 | 2020 | 96 | |
AK134183.1 | - | 21 - 22 | 2180 | 96 | ||
AK146880.1 | - | 21 - 22 | 2180 | 96 | ||
BC003988.1 | - | 11 - 12 | 1134 | 96 | ||
BC005719.1 | - | 16 - 17 | 1790 | 96 | ||
BC023854.1 | - | 21 - 22 | 2170 | 5346780 | 96 | |
BC031899.1 | - | 21 - 22 | 2170 | 5362121 | 96 | |
BC042419.1 | - | 19 - 20 | 5421 | 96 | ||
BC043058.1 | - | 21 - 22 | 2122 | 6419695 | 96 | |
BC054729.1 | - | 21 - 22 | 2173 | 6419758 | 96 | |
BC054820.1 | - | 19 - 20 | 4899 | 96 | ||
BC060985.1 | - | 20 - 21 | 2089 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |