Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029838.2 | NP_001025009.1 | 8 - 9 | 996 | 73 | |
NM_148950.3 | NP_683752.2 | 7 - 8 | 891 | 73 | ||
GenBank mRNA | AB086817.1 | - | 5 - 6 | 717 | 73 | |
AF487460.1 | - | 6 - 7 | 797 | 73 | ||
AK082952.1 | - | 8 - 9 | 941 | 73 | ||
BC050865.1 | - | 7 - 8 | 825 | 6332968 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |