Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clock | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289826.1 | NP_001276755.1 | 4 - 5 | 393 | 134 | |
NM_007715.6 | NP_031741.1 | 4 - 5 | 436 | 134 | ||
GenBank mRNA | AF000998.1 | - | 4 - 5 | 436 | 134 | |
AK030352.1 | - | 4 - 5 | 358 | 134 | ||
AK040337.1 | - | 4 - 5 | 374 | 134 | ||
AK041303.1 | - | 4 - 5 | 374 | 134 | ||
AK129118.1 | - | 4 - 5 | 393 | 134 | ||
AK158765.1 | - | 4 - 5 | 374 | 134 | ||
BC129886.1 | - | 2 - 3 | 145 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |