Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007800.1 | NP_031826.1 | 2 - 3 | 342 | 62 | |
GenBank mRNA | BC125511.1 | - | 2 - 3 | 257 | 40130026 | 62 |
BC125513.1 | - | 2 - 3 | 257 | 40130028 | 62 | |
X78544.1 | - | 2 - 3 | 342 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |