Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007842.2 | NP_031868.2 | 20 - 21 | 2511 | 84 | |
GenBank mRNA | U91922.1 | - | 20 - 21 | 2468 | 84 | |
U92080.1 | - | 9 - 10 | 1206 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |