Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008230.6 | NP_032256.3 | 8 - 9 | 1082 | 65 | |
GenBank mRNA | AF109137.1 | - | 8 - 9 | 992 | 65 | |
AK076384.1 | - | 7 - 8 | 3054 | 65 | ||
AK088545.1 | - | 8 - 9 | 1069 | 65 | ||
AK133455.1 | - | 8 - 9 | 1083 | 65 | ||
AK150168.1 | - | 8 - 9 | 1070 | 65 | ||
AK153104.1 | - | 8 - 9 | 1070 | 65 | ||
BC052833.1 | - | 8 - 9 | 1067 | 30044098 | 65 | |
X57437.1 | - | 8 - 9 | 1047 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |