Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 8 - 9 | 1077 | 78 | |
GenBank mRNA | AF022908.1 | - | 8 - 9 | 1039 | 78 | |
AK029876.1 | - | 8 - 9 | 1078 | 78 | ||
AK150015.1 | - | 8 - 9 | 1058 | 78 | ||
AK152781.1 | - | 8 - 9 | 1058 | 78 | ||
AK162656.1 | - | 2 - 3 | 3016 | 78 | ||
BC090617.1 | - | 8 - 9 | 1048 | 30943198 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |