Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009564.2 | NP_033590.2 | 2 - 3 | 710 | 68 | |
GenBank mRNA | AK010444.1 | - | 2 - 3 | 412 | 68 | |
AK146036.1 | - | 2 - 3 | 396 | 68 | ||
BC004695.1 | - | 392 | 3498906 | 68 | ||
CB182814.2 | - | 1 - 2 | 154 | 68 | ||
CX205658.1 | - | 2 - 3 | 385 | 68 | ||
U49046.1 | - | 2 - 3 | 382 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |