Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010531.1 | NP_034661.1 | 2 - 3 | 118 | 57 | |
GenBank mRNA | AF110803.1 | - | 1 - 2 | 45 | 57 | |
AF122907.1 | - | 2 - 3 | 118 | 57 | ||
AK003370.1 | - | 2 - 3 | 134 | 57 | ||
AK008452.1 | - | 1 - 2 | 49 | 57 | ||
AK009721.1 | - | 1 - 2 | 45 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |