Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004961.1 | - | 16 - 17 | 1978 | 110 | |
AK128947.1 | - | 16 - 17 | 2005 | 110 | ||
AK158861.1 | - | 16 - 17 | 1977 | 110 | ||
AK158935.1 | - | 17 - 18 | 1893 | 110 | ||
CA977198.1 | - | 2 - 3 | 299 | 110 | ||
X60831.1 | - | 16 - 17 | 1956 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |