Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013675.3 | NP_038703.3 | 6 - 7 | 1316 | 62 | |
GenBank mRNA | AK147299.1 | - | 6 - 7 | 1317 | 62 | |
AK147684.1 | - | 6 - 7 | 909 | 62 | ||
AK147704.1 | - | 6 - 7 | 909 | 62 | ||
BC129806.1 | - | 5 - 6 | 649 | 40087684 | 62 | |
BC150783.1 | - | 5 - 6 | 682 | 9087694 | 62 | |
S66283.1 | - | 6 - 7 | 792 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |