Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038700.2 | NP_001033789.1 | 8 - 9 | 1000 | 73 | |
NM_001177648.1 | NP_001171119.1 | 8 - 9 | 1000 | 73 | ||
NM_001177649.1 | NP_001171120.1 | 8 - 9 | 1000 | 73 | ||
NM_001177650.1 | NP_001171121.1 | 6 - 7 | 697 | 73 | ||
NM_019406.3 | NP_062279.1 | 8 - 9 | 1000 | 73 | ||
GenBank mRNA | AK052652.1 | - | 2 - 3 | 292 | 73 | |
AK122308.1 | - | 8 - 9 | 1000 | 73 | ||
AK143512.1 | - | 6 - 7 | 698 | 73 | ||
AK157135.1 | - | 8 - 9 | 991 | 73 | ||
AK170497.1 | - | 8 - 9 | 950 | 73 | ||
AK172100.1 | - | 8 - 9 | 944 | 73 | ||
BC003867.1 | - | 8 - 9 | 931 | 3490113 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |