Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081695.2 | NP_001075164.1 | 5 - 6 | 541 | 65 | |
NM_001284197.1 | NP_001271126.1 | 3 - 4 | 382 | 65 | ||
NM_019448.4 | NP_062321.1 | 4 - 5 | 395 | 65 | ||
GenBank mRNA | AF220524.1 | - | 3 - 4 | 326 | 65 | |
AJ404467.1 | - | 3 - 4 | 367 | 65 | ||
AK010434.1 | - | 4 - 5 | 396 | 65 | ||
AV473569.1 | - | 3 - 4 | 382 | 65 | ||
BC083147.1 | - | 3 - 4 | 380 | 30447906 | 65 | |
EF051624.1 | - | 5 - 6 | 541 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |