Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697011 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697010 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697006 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2027836 |
BC065141.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2027836 |
NM_020275.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242645 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
590912 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
53970 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
108428 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924835 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
565927 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55785 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514689 |
AK035950.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600102 |
AK028258.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600102 |
AK155456.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600102 |
BC057893.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600102 |
NM_173033.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532063 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142357 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96190 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130260 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P174396 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2027836 |
BC065141.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2027836 |
NM_020275.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P10134 |
BC065141.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P10134 |
AK080069.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P10134 |
NM_020275.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P351638 |
BC065141.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P351638 |
AK145730.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P351638 |
AK154993.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P351638 |
AK016138.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P351638 |
AK050753.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P389028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697005 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697006 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696999 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697002 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696998 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696997 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697013 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697007 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697016 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697011 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697015 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696995 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696980 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696990 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696993 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696996 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696978 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29696977 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697021 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697004 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29697019 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08152579 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130260 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08152596 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P174396 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P10134 |
BC065141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P10134 |
AK080069.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P10134 |
NM_020275.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P351638 |
BC065141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P351638 |
AK145730.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P351638 |
AK154993.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P351638 |
AK016138.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P351638 |
AK050753.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P389028 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135855 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P174396 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130260 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08152596 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11762079 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11433283 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11762081 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11474387 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697006 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697010 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29697011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29696990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29697035 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696995 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11181492 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11295164 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697019 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11129120 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11404370 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697026 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696977 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697011 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697025 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11683045 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696969 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696974 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697005 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697004 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697030 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697029 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11562865 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697002 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697048 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697013 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11821547 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11762076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697016 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11181488 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11295184 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11180891 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697015 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696990 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697012 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11232024 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11316483 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11125354 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697023 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696967 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697047 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696997 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696988 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29697039 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29696980 |
- |
- |