Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013771.mm.1 |
TC1600001679.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013769.mm.1 |
TC1600001679.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013770.mm.1 |
TC1600001679.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013768.mm.1 |
TC1600001679.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013772.mm.1 |
TC1600001679.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021495.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021495.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021496.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021496.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021497.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
NM_021497.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013773.mm.1 |
TC1600001679.mm.1 |
MTA TC |