Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022420.2 | NP_071865.1 | 3 - 4 | 1356 | 69 | |
GenBank mRNA | AB030197.1 | - | 2 - 3 | 739 | 69 | |
AF378831.1 | - | 3 - 4 | 1308 | 69 | ||
AK032047.1 | - | 3 - 4 | 1217 | 69 | ||
BC020004.1 | - | 3 - 4 | 1307 | 3499841 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |