Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023637.3 | NP_076126.2 | 8 - 9 | 860 | 104 | |
GenBank mRNA | AB029949.1 | - | 8 - 9 | 832 | 104 | |
AK010491.1 | - | 8 - 9 | 838 | 104 | ||
AK087606.1 | - | 8 - 9 | 827 | 104 | ||
AK140760.1 | - | 8 - 9 | 827 | 104 | ||
AK178578.1 | - | 2 - 3 | 134 | 104 | ||
BC079664.1 | - | 8 - 9 | 837 | 30357834 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |