Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163733.1 | NP_001157205.1 | 2 - 3 | 126 | 103 | |
NM_001291182.1 | NP_001278111.1 | 2 - 3 | 126 | 103 | ||
NM_023649.2 | NP_076138.2 | 2 - 3 | 126 | 103 | ||
NM_153677.2 | NP_710143.2 | 2 - 3 | 126 | 103 | ||
GenBank mRNA | AF228924.1 | - | 2 - 3 | 126 | 103 | |
AF228925.1 | - | 2 - 3 | 126 | 103 | ||
AY103465.1 | - | 2 - 3 | 126 | 103 | ||
AY246556.1 | - | 2 - 3 | 100 | 103 | ||
BC010819.1 | - | 2 - 3 | 128 | 4217086 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |