Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024245.4 | NP_077207.3 | 4 - 5 | 493 | 102 | |
GenBank mRNA | AK013939.1 | - | 4 - 5 | 431 | 102 | |
AK030771.1 | - | 4 - 5 | 470 | 102 | ||
BC003363.1 | - | 4 - 5 | 466 | 102 | ||
BC047273.1 | - | 4 - 5 | 451 | 6440797 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |