Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024499.1 | NP_078775.1 | 9 - 10 | 807 | 74 | |
GenBank mRNA | AK039966.1 | - | 8 - 9 | 802 | 74 | |
AK051005.1 | - | 9 - 10 | 806 | 74 | ||
AK079168.1 | - | 9 - 10 | 817 | 74 | ||
AK165557.1 | - | 9 - 10 | 831 | 74 | ||
BC003836.1 | - | 9 - 10 | 807 | 3486148 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |