Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025454.2 | NP_079730.1 | 2 - 3 | 139 | 92 | |
GenBank mRNA | AK006421.1 | - | 2 - 3 | 238 | 92 | |
AK007536.1 | - | 2 - 3 | 130 | 92 | ||
AK009312.1 | - | 2 - 3 | 123 | 92 | ||
AK028010.1 | - | 2 - 3 | 117 | 92 | ||
AK028397.1 | - | 2 - 3 | 140 | 92 | ||
AK145695.1 | - | 2 - 3 | 112 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |