Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290761.1 | NP_001277690.1 | 1 - 2 | 465 | 67 | |
NM_001290762.1 | NP_001277691.1 | 1 - 2 | 465 | 67 | ||
NM_001290763.1 | NP_001277692.1 | 1 - 2 | 465 | 67 | ||
NM_026393.2 | NP_080669.1 | 1 - 2 | 465 | 67 | ||
GenBank mRNA | AK012340.1 | - | 1 - 2 | 221 | 67 | |
AK078389.1 | - | 1 - 2 | 221 | 67 | ||
AK146521.1 | - | 220 | 67 | |||
BC030039.1 | - | 1 - 2 | 168 | 1313824 | 67 | |
BX527978.1 | - | 1 - 2 | 466 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |