Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163746.1 | NP_001157218.1 | 5 - 6 | 705 | 105 | |
NM_028352.4 | NP_082628.3 | 5 - 6 | 705 | 105 | ||
GenBank mRNA | AK013402.1 | - | 5 - 6 | 688 | 105 | |
AK028066.1 | - | 5 - 6 | 695 | 105 | ||
AK032776.1 | - | 5 - 6 | 657 | 105 | ||
AK036561.1 | - | 5 - 6 | 691 | 105 | ||
AK043547.1 | - | 5 - 6 | 706 | 105 | ||
AK049337.1 | - | 5 - 6 | 657 | 105 | ||
AK049758.1 | - | 5 - 6 | 692 | 105 | ||
AK051706.1 | - | 5 - 6 | 645 | 105 | ||
AK153501.1 | - | 4 - 5 | 525 | 105 | ||
AK160913.1 | - | 5 - 6 | 706 | 105 | ||
AK165513.1 | - | 5 - 6 | 690 | 105 | ||
AK169082.1 | - | 5 - 6 | 670 | 105 | ||
AK169141.1 | - | 5 - 6 | 664 | 105 | ||
AK169787.1 | - | 5 - 6 | 679 | 105 | ||
BC138700.1 | - | 5 - 6 | 630 | 8861722 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |