Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028860.2 | NP_083136.2 | 13 - 14 | 1800 | 90 | |
GenBank mRNA | AK141138.1 | - | 13 - 14 | 1780 | 90 | |
BC032166.1 | - | 13 - 14 | 1783 | 5352919 | 90 | |
BC081544.1 | - | 13 - 14 | 1784 | 3672099 | 90 | |
CB527330.1 | - | 4 - 5 | 564 | 90 | ||
CF532066.1 | - | 1 - 2 | 142 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |