Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002238.2 | NP_001002238.1 | 17 - 18 | 2391 | 74 | |
NM_001002240.2 | NP_001002240.1 | 17 - 18 | 2364 | 74 | ||
NM_001002241.2 | NP_001002241.1 | 16 - 17 | 2664 | 74 | ||
NM_031387.3 | NP_113564.2 | 17 - 18 | 2524 | 74 | ||
GenBank mRNA | AB067571.1 | - | 16 - 17 | 2345 | 74 | |
AB183526.1 | - | 17 - 18 | 2391 | 74 | ||
AB183527.1 | - | 17 - 18 | 2364 | 74 | ||
AB183528.1 | - | 16 - 17 | 2664 | 74 | ||
AB183529.1 | - | 16 - 17 | 2228 | 74 | ||
AF285591.1 | - | 13 - 14 | 1718 | 74 | ||
AK029806.1 | - | 17 - 18 | 2525 | 74 | ||
BC129954.1 | - | 12 - 13 | 1650 | 40081321 | 74 | |
BC129955.1 | - | 12 - 13 | 1650 | 40081323 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |