Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053253.3 | NP_444483.2 | 9 - 10 | 1150 | 101 | |
GenBank mRNA | AF123386.1 | - | 7 - 8 | 781 | 101 | |
AK132332.1 | - | 9 - 10 | 1159 | 101 | ||
BC052357.1 | - | 9 - 10 | 1150 | 6772144 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |