Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080467.3 | NP_536715.3 | 8 - 9 | 997 | 83 | |
GenBank mRNA | AB050903.1 | - | 7 - 8 | 723 | 83 | |
AF326316.1 | - | 8 - 9 | 791 | 83 | ||
AF435090.1 | - | 8 - 9 | 985 | 83 | ||
AK128892.1 | - | 8 - 9 | 997 | 83 | ||
BC046979.1 | - | 9 - 10 | 1019 | 6315485 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |