Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100002931.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002931.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002931.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002931.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002931.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002916.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002916.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002916.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002916.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002916.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002918.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002918.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002918.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002918.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002918.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002926.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002926.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002926.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002932.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002932.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002932.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002932.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002932.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002933.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002933.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002933.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002933.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002933.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002917.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002917.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002917.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002917.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002917.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002924.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002924.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002924.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002927.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002927.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002927.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002927.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002927.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002921.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002921.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002921.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002921.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002921.mm.1 |
TC1100000344.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002920.mm.1 |
NM_001271347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002920.mm.1 |
NM_001271348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002920.mm.1 |
NM_001271349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002920.mm.1 |
NM_134015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002920.mm.1 |
TC1100000344.mm.1 |
MTA TC |