Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138304.2 | NP_612177.1 | 3 - 4 | 229 | 63 | |
GenBank mRNA | AK008039.1 | - | 3 - 4 | 215 | 63 | |
AK146656.1 | - | 3 - 4 | 202 | 63 | ||
AY061807.1 | - | 3 - 4 | 229 | 63 | ||
BC013541.1 | - | 3 - 4 | 207 | 4241711 | 63 | |
CO039047.1 | - | 2 - 3 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |