Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199349.1 | NP_001186278.1 | 5 - 6 | 617 | 102 | |
NM_138309.3 | NP_612182.1 | 4 - 5 | 548 | 102 | ||
GenBank mRNA | AK080225.1 | - | 5 - 6 | 373 | 102 | |
AK166275.1 | - | 5 - 6 | 368 | 102 | ||
AY078163.1 | - | 4 - 5 | 344 | 102 | ||
BC031736.1 | - | 4 - 5 | 323 | 4013657 | 102 | |
EF516990.1 | - | 5 - 6 | 375 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |