Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26373435 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
195073 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312780 |
AF007116.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312780 |
AK034475.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312780 |
BC119606.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312780 |
BC119607.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312780 |
NM_011800.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607388 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
673820 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260969 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84698 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139153 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373449 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373463 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373440 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373435 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373461 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373459 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373458 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373450 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373456 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26373439 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139153 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06730221 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
NM_144543.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
AK077639.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
BC016594.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
BE953030.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323620 |
AK163860.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138143 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139153 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07775836 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07775831 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08005859 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26373435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26373479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07737719 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373463 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373461 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373467 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373444 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373458 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07536261 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373482 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07765059 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373480 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07536262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08098814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373449 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07460362 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07689452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373450 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373440 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26373437 |
- |
- |