Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199988.1 | NP_001186917.1 | 1 - 2 | 1180 | 84 | |
NM_144927.3 | NP_659176.2 | 1 - 2 | 1180 | 84 | ||
NR_103719.1 | 1 - 2 | 1180 | 84 | |||
GenBank mRNA | AK045392.1 | - | 1 - 2 | 1185 | 84 | |
AK046031.1 | - | 1 - 2 | 1170 | 84 | ||
AK084242.1 | - | 1 - 2 | 1170 | 84 | ||
AK139425.1 | - | 1 - 2 | 1157 | 84 | ||
AK214154.1 | - | 1 - 2 | 65 | 84 | ||
AK219102.1 | - | 1 - 2 | 66 | 84 | ||
BC019943.1 | - | 1 - 2 | 883 | 4017819 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |