Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145139.2 | NP_660121.2 | 2 - 3 | 121 | 59 | |
GenBank mRNA | AJ310346.1 | - | 2 - 3 | 131 | 59 | |
AK032936.1 | - | 2 - 3 | 109 | 59 | ||
AK075584.1 | - | 2 - 3 | 127 | 59 | ||
AK150895.1 | - | 2 - 3 | 116 | 59 | ||
AK151175.1 | - | 2 - 3 | 116 | 59 | ||
AK167640.1 | - | 2 - 3 | 116 | 59 | ||
AK168009.1 | - | 2 - 3 | 119 | 59 | ||
BC024463.1 | - | 2 - 3 | 121 | 4975621 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |