Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145228.2 | NP_660263.1 | 4 - 5 | 957 | 65 | |
GenBank mRNA | AB067530.1 | - | 4 - 5 | 957 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |