Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145542.3 | NP_663517.2 | 6 - 7 | 946 | 111 | |
GenBank mRNA | AB092504.1 | - | 6 - 7 | 675 | 111 | |
BC018218.1 | - | 6 - 7 | 775 | 4007102 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |