Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082536.1 | NP_001076005.1 | 8 - 9 | 1195 | 87 | |
NM_153049.3 | NP_694629.2 | 7 - 8 | 957 | 87 | ||
GenBank mRNA | AF385582.1 | - | 7 - 8 | 728 | 87 | |
AF532597.1 | - | 8 - 9 | 1079 | 87 | ||
AK044188.1 | - | 8 - 9 | 1188 | 87 | ||
AK089416.1 | - | 6 - 7 | 787 | 87 | ||
AK154561.1 | - | 8 - 9 | 1175 | 87 | ||
AK155937.1 | - | 7 - 8 | 941 | 87 | ||
AK157985.1 | - | 6 - 7 | 806 | 87 | ||
BC050941.1 | - | 8 - 9 | 1088 | 6821370 | 87 | |
BC054801.1 | - | 8 - 9 | 1131 | 87 | ||
CA318154.1 | - | 2 - 3 | 222 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |