Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153057.4 | NP_694697.3 | 23 - 24 | 2871 | 77 | |
GenBank mRNA | AK030584.1 | - | 23 - 24 | 2838 | 77 | |
AK035957.1 | - | 23 - 24 | 2843 | 77 | ||
AK080754.1 | - | 23 - 24 | 2734 | 77 | ||
AK082495.1 | - | 23 - 24 | 2750 | 77 | ||
AK082963.1 | - | 23 - 24 | 2730 | 77 | ||
AK142338.1 | - | 23 - 24 | 2841 | 77 | ||
AK150107.1 | - | 23 - 24 | 2826 | 77 | ||
AK165643.1 | - | 23 - 24 | 2734 | 77 | ||
AK166764.1 | - | 8 - 9 | 881 | 77 | ||
AK195057.1 | - | 94 | 77 | |||
AK198231.1 | - | 1 - 2 | 94 | 77 | ||
AK203299.1 | - | 2 - 3 | 132 | 77 | ||
BC022138.1 | - | 22 - 23 | 2587 | 77 | ||
BC024503.1 | - | 1 - 2 | 114 | 77 | ||
BC025133.1 | - | 6 - 7 | 750 | 77 | ||
BC033923.1 | - | 12 - 13 | 1455 | 5099899 | 77 | |
BC072630.1 | - | 23 - 24 | 2817 | 6833516 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |