Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242427.1 | NP_001229356.1 | 7 - 8 | 1413 | 61 | |
NM_153062.2 | NP_694702.1 | 7 - 8 | 993 | 61 | ||
GenBank mRNA | AK033656.1 | - | 7 - 8 | 882 | 61 | |
AK143636.1 | - | 7 - 8 | 1419 | 61 | ||
AK153706.1 | - | 7 - 8 | 1416 | 61 | ||
AK163990.1 | - | 6 - 7 | 595 | 61 | ||
BC027294.1 | - | 7 - 8 | 993 | 3985469 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |