Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2029902 |
AY057988.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2029902 |
BC141240.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2029902 |
AK031227.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2029902 |
AK039787.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2029902 |
NM_181584.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408844 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
107446 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
661844 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395549 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
98594 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
561085 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
378775 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
662305 |
XM_001472240.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121858 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831247 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515715 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
17722 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842501 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
109740 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
778082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
753391 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
234865 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
185304 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
171031 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731240 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541191 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
466341 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
247381 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
345856 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
525268 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
942721 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911353 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
161823 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651832 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
298105 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773047 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
458528 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516709 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
677846 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
832856 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
146538 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154500 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
834701 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3733 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
677433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510795 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
25967 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11335 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
161273 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
118001 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
87200 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
93845 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
90818 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151749 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
122049 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140991 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P117594 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2029902 |
AY057988.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2029902 |
BC141240.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2029902 |
AK031227.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2029902 |
AK039787.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2029902 |
NM_181584.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470527 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470528 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470533 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470529 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470540 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470512 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470523 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470536 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470542 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470519 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470520 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470510 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470508 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470534 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470524 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470539 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470545 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470537 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470544 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470526 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470518 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470522 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470531 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470509 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470516 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32470514 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
AY057988.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
BC141240.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
AK031227.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
AK039787.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P433817 |
NM_181584.4 |
mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14897857 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15017170 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14897858 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14897855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32470544 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470515 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470534 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470520 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470524 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470527 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470522 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470531 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470512 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470518 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470528 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470529 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470519 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15017170 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470508 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470526 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470523 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470509 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470516 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470533 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32470510 |
- |
- |