Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153077.2 | NP_694717.1 | 1 - 2 | 52 | 89 | |
GenBank mRNA | AY071844.1 | - | 1 - 2 | 52 | 89 | |
BC107311.1 | - | 1 - 2 | 44 | 40053322 | 89 | |
BC107312.1 | - | 1 - 2 | 48 | 40053325 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |