Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252515.1 | NP_001239444.1 | 21 - 22 | 3713 | 59 | |
NM_153078.4 | NP_694718.4 | 21 - 22 | 3638 | 59 | ||
GenBank mRNA | AF424697.1 | - | 20 - 21 | 3488 | 59 | |
AK173055.1 | - | 21 - 22 | 3714 | 59 | ||
BC037023.1 | - | 8 - 9 | 1265 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |