Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114665.2 | NP_001108137.1 | 14 - 15 | 1666 | 91 | |
NM_153118.3 | NP_694758.2 | 12 - 13 | 1492 | 91 | ||
GenBank mRNA | BC034510.1 | - | 7 - 8 | 940 | 4481144 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |