Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153128.2 | NP_694768.1 | 3 - 4 | 507 | 70 | |
GenBank mRNA | AK034170.1 | - | 3 - 4 | 495 | 70 | |
BC034514.1 | - | 3 - 4 | 507 | 3157064 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |