Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153131.3 | NP_694771.1 | 13 - 14 | 2416 | 66 | |
GenBank mRNA | AJ487852.1 | - | 13 - 14 | 2417 | 66 | |
AK122575.1 | - | 9 - 10 | 2738 | 66 | ||
BC058084.1 | - | 12 - 13 | 2269 | 6813463 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |