Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162868.1 | NP_001156340.1 | 7 - 8 | 2272 | 59 | |
NM_001162869.1 | NP_001156341.1 | 8 - 9 | 2407 | 59 | ||
NM_153140.2 | NP_694780.1 | 5 - 6 | 663 | 59 | ||
GenBank mRNA | AB093257.1 | - | 8 - 9 | 1848 | 59 | |
AK083658.1 | - | 7 - 8 | 1956 | 59 | ||
AK085516.1 | - | 5 - 6 | 664 | 59 | ||
AK128985.1 | - | 5 - 6 | 663 | 59 | ||
BC023364.1 | - | 5 - 6 | 654 | 59 | ||
BC037132.1 | - | 5 - 6 | 703 | 59 | ||
CF170100.1 | - | 5 - 6 | 562 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |