Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013045.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013045.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013045.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013039.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013039.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013039.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013041.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013041.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013041.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013040.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013040.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013040.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013043.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013043.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013043.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013042.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013042.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013042.mm.1 |
TC1700001458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013044.mm.1 |
NM_153151.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013044.mm.1 |
NM_153151.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013044.mm.1 |
TC1700001458.mm.1 |
MTA TC |