Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153176.4 | NP_694816.3 | 6 - 7 | 884 | 63 | |
GenBank mRNA | AF512565.1 | - | 6 - 7 | 916 | 63 | |
AF547215.1 | - | 6 - 7 | 1213 | 63 | ||
AK145540.1 | - | 6 - 7 | 885 | 63 | ||
AK171647.1 | - | 5 - 6 | 783 | 63 | ||
BC024986.1 | - | 4 - 5 | 539 | 63 | ||
BC051051.1 | - | 6 - 7 | 818 | 63 | ||
BC055488.1 | - | 729 | 63 | |||
BC138141.1 | - | 6 - 7 | 1111 | 8861160 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |